Tetrahydrofuran-3-ylmethanol ≥95%

Supplier: Matrix Scientific

Synonyms: Oxolan-3-ylmethanol, (Tetrahydrofuran-3-yl)methanol

068672-1G 068672-25G
101937-180EA 106.15 USD
101937-180 102663-836
Tetrahydrofuran-3-ylmethanol ≥95%
Tetrahydrofuran-3-ylmethanol

Hazard: Irritant

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
MDL Number: MFCD00075198
CAS Number: 15833-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR