1H-Pyrrolo[2,3-b]pyridine-3-methanamine ≥95%

Supplier: Matrix Scientific

068660-500MG 068660-1G
101937-132EA 984.62 USD
101937-132 101937-134
1H-Pyrrolo[2,3-b]pyridine-3-methanamine ≥95%
1H-Pyrrolo[2,3-b]pyridine-3-methanamine

Hazard: Irritant

Formula: C₈H₉N₃
MDL Number: MFCD12964704
CAS Number: 933691-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR