1H-Pyrrolo[2,3-b]pyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Azaindole-3-carbonitrile, 3-Cyano-1H-pyrrolo[2,3-b]pyridine,3-Cyano-7-azaindole

068659-1G 068659-5G
101937-128EA 204.62 USD
101937-128 101937-130
1H-Pyrrolo[2,3-b]pyridine-3-carbonitrile ≥95%
1H-Pyrrolo[2,3-b]pyridine-3-carbonitrile

Hazard: Irritant

Formula: C₈H₅N₃
MW: 143.15 g/mol
MDL Number: MFCD07778359
CAS Number: 4414-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR