1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone ≥95%

Supplier: Matrix Scientific

068658-250MG 068658-500MG
101937-124EA 640 USD
101937-124 101937-126
1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone ≥95%
1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD07636699
CAS Number: 453562-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR