1H-Pyrrolo[2,3-b]pyridine-5-methanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Aminomethyl-7-azaindole

068656-250MG 068656-500MG 068656-1G
101937-112EA 1163.08 USD
101937-112 101937-114 101937-116
1H-Pyrrolo[2,3-b]pyridine-5-methanamine ≥95%
1H-Pyrrolo[2,3-b]pyridine-5-methanamine

Hazard: Irritant

Formula: C₈H₉N₃
MW: 147.18 g/mol
MDL Number: MFCD09475794
CAS Number: 267876-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR