7-Azaindole-5-boronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (1H-Pyrrolo[2,3-b]pyridin-5-yl)boronic acid, {1H-pyrrolo[2,3-b]pyridin-5-yl}boronic acid

068655-250MG 068655-500MG
101937-108EA 181.54 USD
101937-108 101937-110
7-Azaindole-5-boronic acid ≥95%
7-Azaindole-5-boronic acid

Hazard: Irritant

Formula: C₇H₇BN₂O₂
MW: 161.96 g/mol
MDL Number: MFCD08234881
CAS Number: 944059-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR