Methyl-3-hydroxycyclobutanecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-hydroxycyclobutanecarboxylate, Methyl 3-hydroxycyclobutane-1-carboxylate
068649-1G
068649-5G
068649-10G
101937-074EA
161.54
USD
101937-074
101937-076
101937-078
Methyl-3-hydroxycyclobutanecarboxylate ≥95%
Methyl-3-hydroxycyclobutanecarboxylate
Hazard: Irritant
Formula:
C₆H₁₀O₃ MW: 130.14 g/mol |
MDL Number:
MFCD00512755 CAS Number: 4934-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...