3-Aminocyclobutanemethanol hydrochloride ≥95%

Supplier: Matrix Scientific

068645-250MG 068645-500MG 068645-1G
101937-052EA 307.69 USD
101937-052 101937-054 101937-056
3-Aminocyclobutanemethanol hydrochloride ≥95%
3-Aminocyclobutanemethanol hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MW: 137.61 g/mol
MDL Number: MFCD16659926
CAS Number: 130369-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR