3-Cyclobutyl-3-oxo-propionitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Cyclobutyl-3-oxopropanenitrile, b-Oxo-cyclobutanepropanenitrile

068643-1G 068643-5G
101937-044EA 615.38 USD
101937-044 101937-046
3-Cyclobutyl-3-oxo-propionitrile ≥95%
3-Cyclobutyl-3-oxo-propionitrile

Hazard: Irritant

Formula: C₇H₉NO
MW: 123.16 g/mol
MDL Number: MFCD07787182
CAS Number: 118431-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR