1-Boc-3-methoxycarbonylmethyleneazetidine ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(2-methoxy-2-oxoethylidene)azetidine-1-carboxylate

068639-250MG 068639-500MG
101937-026EA 640 USD
101937-026 101937-028
1-Boc-3-methoxycarbonylmethyleneazetidine ≥97%
1-Boc-3-methoxycarbonylmethyleneazetidine

Hazard: Irritant

Formula: C₁₁H₁₇NO₄
MW: 227.26 g/mol
MDL Number: MFCD13190429
CAS Number: 1105662-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR