3-Methylthioazetidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Azetidine-3-thiol hydrochloride

068638-250MG 068638-500MG
101937-022EA 727.69 USD
101937-022 101937-024
3-Methylthioazetidine hydrochloride ≥95%
3-Methylthioazetidine hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNS
MDL Number: MFCD17015916
CAS Number: 179337-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR