O-1-tert-Butyl-O-3-methyl-(3R)-pyrrolidine-1,3-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-Methyl 1-Boc-pyrrolidine-3-carboxylate, (R)-1-tert-Butyl 3-methyl pyrrolidine-1,3-dicarboxylate

068637-1G 068637-5G
101937-018EA 1861.54 USD
101937-018 101937-020
O-1-tert-Butyl-O-3-methyl-(3R)-pyrrolidine-1,3-dicarboxylate ≥95%
O-1-tert-Butyl-O-3-methyl-(3R)-pyrrolidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD09954284
CAS Number: 441717-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR