1-Methyl-4-(hydroxyethyl)piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1-Methylpiperidino)ethanol, 2-(1-Methylpiperidin-4-yl)ethanol

068636-1G 068636-5G
101937-014EA 230.77 USD
101937-014 101937-016
1-Methyl-4-(hydroxyethyl)piperidine ≥95%
1-Methyl-4-(hydroxyethyl)piperidine

Hazard: Irritant

Formula: C₈H₁₇NO
MW: 143.23 g/mol
MDL Number: MFCD13188565
CAS Number: 21156-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR