tert-Butyl-3-(2-hydroxyethyl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(2-hydroxyethyl)azetidine-1-carboxylate

068634-250MG 068634-500MG
101937-010EA 524.62 USD
101937-010 101937-012
tert-Butyl-3-(2-hydroxyethyl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(2-hydroxyethyl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD17015912
CAS Number: 152537-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR