(S)-(-)-1-Methyl-2-aminomethylpyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(1-Methylpyrrolidin-2-yl)methanamine

068633-250MG 068633-500MG
101937-006EA 330.77 USD
101937-006 101937-008
(S)-(-)-1-Methyl-2-aminomethylpyrrolidine ≥95%
(S)-(-)-1-Methyl-2-aminomethylpyrrolidine

Hazard: Irritant

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD11106680
CAS Number: 66411-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR