(R)-Pyrrolidine-3-carboxamide hydrochloride ≥95%
Supplier: Matrix Scientific
068632-250MG
068632-500MG
101937-002EA
520
USD
101937-002
101937-004
(R)-Pyrrolidine-3-carboxamide hydrochloride ≥95%
(R)-Pyrrolidine-3-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₅H₁₁ClN₂O |
MDL Number:
MFCD17171383 CAS Number: 1273577-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...