3-Boc-aminomethylpyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Boc-aminomethyl-pyrrolidine

068630-1G 068630-5G
101936-994EA 84.62 USD
101936-994 101936-996
3-Boc-aminomethylpyrrolidine ≥95%
3-Boc-aminomethylpyrrolidine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.2780 g/mol
MDL Number: MFCD04973997
CAS Number: 149366-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR