(R)-3-(Methylsulfonyl)pyrrolidine hydrochloride ≥95%

Supplier: Matrix Scientific

068625-250MG 068625-500MG 068625-1G
101936-972EA 292.31 USD
101936-972 101936-974 101936-976
(R)-3-(Methylsulfonyl)pyrrolidine hydrochloride ≥95%
(R)-3-(Methylsulfonyl)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO₂S
MDL Number: MFCD22416364
CAS Number: 1392745-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR