(S)-1-Methyl-3-pyrrolidinol ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-3-Hydroxy-1-methylpyrrolidine, (S)-(+)-1-Methyl-3-pyrrolidinol

068622-1G 068622-5G
101936-960EA 190.77 USD
101936-960 101936-962
(S)-1-Methyl-3-pyrrolidinol ≥95%
(S)-1-Methyl-3-pyrrolidinol

Hazard: Irritant

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD03788748
CAS Number: 104641-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR