3-Amino-1-methylpiperidine dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methylpiperidin-3-amine dihydrochloride

068620-1G 068620-5G
101936-952EA 436.92 USD
101936-952 101936-954
3-Amino-1-methylpiperidine dihydrochloride ≥95%
3-Amino-1-methylpiperidine dihydrochloride

Hazard: Irritant

Formula: C₆H₁₆Cl₂N₂
MW: 187.11 g/mol
MDL Number: MFCD06796280
CAS Number: 894808-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR