Pyrrolidin-3-ol hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (±)-3-Hydroxypyrrolidine hydrochloride

068615-1G 068615-5G 068615-25G
101936-926EA 87.69 USD
101936-926 101936-928 101936-930
Pyrrolidin-3-ol hydrochloride ≥95%
Pyrrolidin-3-ol hydrochloride

Hazard: Irritant

Formula: C4H10ClNO
MW: 123.58 g/mol
MDL Number: MFCD03001754
CAS Number: 86070-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR