Benzyl-3-oxopyrrolidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Benzyloxycarbonyl-3-pyrrolidone, 1-Carbobenzoxy-3-pyrrolidone, 1-Cbz-3-pyrrolidinone, 1-Cbz-3-pyrrolidone, Benzyl 3-oxopyrrolidine-1-carboxylate, 1-Benzyloxycarbonyl-3-pyrrolidinone, 1-Carbobenzoxy-3-pyrrolidinone
068614-5G
068614-25G
101936-922EA
43.08
USD
101936-922
101936-924
Benzyl-3-oxopyrrolidine-1-carboxylate ≥95%
Benzyl-3-oxopyrrolidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₃NO₃ MW: 219.2365 g/mol |
MDL Number:
MFCD03001711 CAS Number: 130312-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...