(±)-1-Boc-3-hydroxypyrrolidine ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 3-hydroxypyrrolidine-1-carboxylate, 3-Hydroxypyrrolidine, N-BOC protected
068613-1G
068613-5G
068613-25G
101936-916EA
32.31
USD
101936-916
101936-918
102833-430
(±)-1-Boc-3-hydroxypyrrolidine ≥95%
(±)-1-Boc-3-hydroxypyrrolidine
Hazard: Irritant
Formula:
C₉H₁₇NO₃ MW: 187.24 g/mol |
MDL Number:
MFCD04038535 CAS Number: 103057-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...