3-Methoxy-3-methylazetidine hydrochloride ≥95%

Supplier: Matrix Scientific

068610-250MG 068610-500MG
101936-904EA 901.54 USD
101936-904 101936-906
3-Methoxy-3-methylazetidine hydrochloride ≥95%
3-Methoxy-3-methylazetidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MDL Number: MFCD11100995
CAS Number: 905843-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR