3,6-Pyridazinedicarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Pyridazine-3,6-dicarboxylic acid

068609-250MG 068609-500MG
101936-900EA 453.85 USD
101936-900 101936-902
3,6-Pyridazinedicarboxylic acid ≥95%
3,6-Pyridazinedicarboxylic acid

Hazard: Irritant

Formula: C₆H₄N₂O₄
MDL Number: MFCD01318693
CAS Number: 57266-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR