4-Chloropyrido[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

068607-250MG 068607-500MG 068607-1G
101936-890EA 353.85 USD
101936-890 101936-892 101936-894
4-Chloropyrido[3,2-d]pyrimidine ≥95%
4-Chloropyrido[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₇H₄ClN₃
MW: 165.5798 g/mol
MDL Number: MFCD10566748
CAS Number: 51674-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR