1H-Indazole-4-boronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1H-indazole, 1H-indazol-4-boronic acid pinacol ester

068603-1G 068603-5G
101936-872EA 458.46 USD
101936-872 101936-874
1H-Indazole-4-boronic acid pinacol ester ≥95%
1H-Indazole-4-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₃H₁₇BN₂O₂
MW: 244.10 g/mol
MDL Number: MFCD08059267
CAS Number: 885618-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR