Thiazole-5-boronic acid pinacol ester ≥95%
Supplier: Matrix Scientific
Synonyms:
Thiazole-5-boronic Acid Pinacol Ester
068602-1G
068602-5G
101936-868EA
369.23
USD
101936-868
101936-870
Thiazole-5-boronic acid pinacol ester ≥95%
Thiazole-5-boronic acid pinacol ester
Hazard: Irritant
Formula:
C₉H₁₄BNO₂S MW: 211.09 g/mol |
MDL Number:
MFCD10697440 CAS Number: 1086111-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...