Methyl-5-aminopicolinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-aminopyridine-2-carboxylate, Methyl 5-aminopicolinate
068601-1G
068601-5G
101936-864EA
116.92
USD
101936-864
101936-866
Methyl-5-aminopicolinate ≥95%
Methyl 5-aminopicolinate
Hazard: Irritant
Formula:
C₇H₈N₂O₂ |
MDL Number:
MFCD07783939 CAS Number: 67515-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...