5-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one ≥95%

Supplier: Matrix Scientific

068597-500MG 068597-1G
101936-846EA 753.85 USD
101936-846 101936-848
5-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one ≥95%
5-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one

Hazard: Irritant

Formula: C₉H₁₀N₂O
MDL Number: MFCD17214360
CAS Number: 1190380-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR