2,3-Dimethyl-6-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-dimethyl-6-nitro-2H-indazole

068596-1G 068596-5G
101936-842EA 69.23 USD
101936-842 101936-844
2,3-Dimethyl-6-nitro-1H-indazole ≥95%
2,3-Dimethyl-6-nitro-1H-indazole

Hazard: Irritant

Formula: C₉H₁₁N₃O₂
MW: 191.1867 g/mol
MDL Number: MFCD10703326
CAS Number: 444731-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR