tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 2,5-dihydro-1H-pyrrole-1-carboxylate, N-Boc-2,5-dihydropyrrole, N-BOC-3-Pyrroline
068594-5G
068594-25G
101936-832EA
30.77
USD
101936-832
101936-834
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥95%
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate
Hazard: Irritant/Refrigerate
Formula:
C₉H₁₅NO₂ MW: 169.224 g/mol Boiling Pt: 208°C Density: 0.981 Flash Pt: 81°C |
MDL Number:
MFCD01863512 CAS Number: 73286-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...