6-Amino-2,3-dimethyl-2H-indazole ≥95%

Supplier: Matrix Scientific

068592-1G 068592-5G
101936-820EA 669.23 USD
101936-820 101936-822
6-Amino-2,3-dimethyl-2H-indazole ≥95%
6-Amino-2,3-dimethyl-2H-indazole

Hazard: Irritant

Formula: C₉H₁₁N₃
MDL Number: MFCD09258896
CAS Number: 444731-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR