cis-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
cis-3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, cis-3-BOC-Aminocyclobutanecarboxylic acid
068590-250MG
068590-500MG
068590-1G
101936-810EA
66.15
USD
101936-810
101936-812
101936-814
cis-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid ≥95%
cis-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₇NO₄ |
MDL Number:
MFCD06657680 CAS Number: 1008773-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...