3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, 3-{[(tert-butoxy)carbonyl]amino}cyclobutane-1-carboxylic acid

068589-500MG 068589-1G
101936-806EA 595.38 USD
101936-806 101936-808
3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid ≥95%
3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD09864181
CAS Number: 946152-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR