trans-3-Aminocyclobutanecarboxylic acid ≥95%

Supplier: Matrix Scientific

068588-1G 068588-5G
101936-802EA 2838.46 USD
101936-802 101936-804
trans-3-Aminocyclobutanecarboxylic acid ≥95%
trans-3-Aminocyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₅H₉NO₂
MDL Number: MFCD06659965
CAS Number: 74307-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR