cis-3-Aminocyclobutanecarboxylic acid ≥95%

Supplier: Matrix Scientific

068587-250MG 068587-500MG
101936-798EA 1221.54 USD
101936-798 101936-800
cis-3-Aminocyclobutanecarboxylic acid ≥95%
cis-3-Aminocyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₅H₉NO₂
MDL Number: MFCD06659964
CAS Number: 74316-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR