3-Amino-1-methylpyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

068585-250MG 068585-500MG
101936-788EA 300 USD
101936-788 101936-790
3-Amino-1-methylpyridin-2(1H)-one ≥95%
3-Amino-1-methylpyridin-2(1H)-one

Hazard: Irritant

Formula: C₆H₈N₂O
MDL Number: MFCD12402373
CAS Number: 33631-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR