5-(Hydrazinomethyl)pyrimidine ≥95%
Supplier: Matrix Scientific
068581-250MG
068581-500MG
068581-1G
101936-774EA
1500
USD
101936-774
101936-776
101936-778
5-(Hydrazinomethyl)pyrimidine ≥95%
5-(Hydrazinomethyl)pyrimidine
Hazard: Irritant
Formula:
C₅H₈N₄ |
MDL Number:
MFCD17015884 CAS Number: 1234616-20-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...