4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

068580-250MG 068580-500MG 068580-1G
101936-768EA 1396.92 USD
101936-768 101936-770 101936-772
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid methyl ester ≥95%
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₈H₆ClN₃O₂
MDL Number: MFCD09702013
CAS Number: 944709-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR