Methyl pyridazine-3-carboxylate ≥95%

Supplier: Matrix Scientific

068579-1G 068579-5G
101936-764EA 129.23 USD
101936-764 101936-766
Methyl pyridazine-3-carboxylate ≥95%
Methyl pyridazine-3-carboxylate

Hazard: Irritant

Formula: C₆H₆N₂O₂
MDL Number: MFCD10698044
CAS Number: 34253-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR