(1R,5S,6S)-N,N-dibenzyl-3-azabicyclo[3.1.0]hexan-6-amine ≥95%

Supplier: Matrix Scientific

068572-250MG 068572-500MG 068572-1G
101936-728EA 553.85 USD
101936-728 101936-730 101936-732
(1R,5S,6S)-N,N-dibenzyl-3-azabicyclo[3.1.0]hexan-6-amine ≥95%
(1R,5S,6S)-N,N-dibenzyl-3-azabicyclo[3.1.0]hexan-6-amine

Hazard: Irritant

Formula: C₁₉H₂₂N₂
MW: 278.4 g/mol
MDL Number: MFCD16556173
CAS Number: 210482-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR