(3S)-3-Methoxypyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-3-Methoxypyrrolidine

068570-1G 068570-5G
101936-720EA 1135.38 USD
101936-720 101936-722
(3S)-3-Methoxypyrrolidine ≥95%
(3S)-3-Methoxypyrrolidine

Hazard: Irritant

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD09953438
CAS Number: 120099-61-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR