(3R)-3-Methoxypyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-3-Methoxypyrrolidine

068569-1G 068569-5G
101936-716EA 1135.38 USD
101936-716 101936-718
(3R)-3-Methoxypyrrolidine ≥95%
(3R)-3-Methoxypyrrolidine

Hazard: Irritant

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD09953439
CAS Number: 120099-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR