3-Methoxypyrrolidine ≥95%

Supplier: Matrix Scientific

068568-250MG 068568-500MG 068568-1G
101936-710EA 407.69 USD
101936-710 101936-712 101936-714
3-Methoxypyrrolidine ≥95%
3-Methoxypyrrolidine

Hazard: Irritant

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD07787006
CAS Number: 62848-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR