tert-Butyl-3-methyleneazetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methylideneazetidine, N-BOC protected, tert-Butyl 3-methyleneazetidine-1-carboxylate

068566-250MG 068566-500MG
101936-706EA 610.77 USD
101936-706 101936-708
tert-Butyl-3-methyleneazetidine-1-carboxylate ≥95%
tert-Butyl-3-methyleneazetidine-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₅NO₂
MW: 169.23 g/mol
MDL Number: MFCD12031229
CAS Number: 934664-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR