(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥95%
Supplier: Matrix Scientific
Synonyms:
(3R)-(+)-3-(Boc-amino)pyrrolidine, (R)-tert-Butyl pyrrolidin-3-ylcarbamate
068565-5G
068565-25G
101936-704EA
113.85
USD
101936-704
102551-714
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥95%
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine
Hazard: Irritant
Formula:
C₉H₁₈N₂O₂ MW: 186.26 g/mol |
MDL Number:
MFCD00143191 CAS Number: 122536-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...