(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-tert-Butyl pyrrolidin-3-ylcarbamate, (3S)-(-)-3-(Boc-amino)pyrrolidine

068564-5G 068564-25G
101936-700EA 66.15 USD
101936-700 102551-712
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥95%
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine

Hazard: Corrosive/Air Sensitive

Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
Melting Pt: 50…54 °C
MDL Number: MFCD00143194
CAS Number: 122536-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR