(S)-(-)-1-Boc-3-aminopyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-1-Boc-3-Aminopyrrolidine, (S)-(-)-1-BOC-3-aminopyrrolidine, tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate

068562-5G 068562-25G
101936-692EA 233.85 USD
101936-692 102551-710
(S)-(-)-1-Boc-3-aminopyrrolidine ≥95%
(S)-(-)-1-Boc-3-aminopyrrolidine

Hazard: Corrosive/Air Sensitive

Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Boiling Pt: 115 °C (4 mmHg)
Density: 1.02
MDL Number: MFCD03419271
CAS Number: 147081-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR