7-(Benzenesulfonyl)-4-chloro-6-iodopyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

068560-250MG 068560-500MG 068560-1G
101936-680EA 669.23 USD
101936-680 101936-682 101936-684
7-(Benzenesulfonyl)-4-chloro-6-iodopyrrolo[2,3-d]pyrimidine ≥95%
7-(Benzenesulfonyl)-4-chloro-6-iodopyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₁₂H₇ClIN₃O₂S
MDL Number: MFCD17015878
CAS Number: 876343-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR